Skip to Content
Merck
All Photos(1)

Key Documents

CH6H8C2C4558

Chem Impex International, Inc.

N-(1-Ethoxyethylidene)2-aminooxyacetic acid N-hydroxysuccinimide ester

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
258.23
MDL number:

mp

53 - 62 C

Iupac Name

ethyl (1E)-N-{2-[(2,5-dioxo-1-pyrrolidinyl)oxy]-2-oxoethoxy}ethanimidoate

Inchi Code

1S/C10H14N2O6/c1-3-16-7(2)11-17-6-10(15)18-12-8(13)4-5-9(12)14/h11H,2-6H2,1H3

InChI key

HRYOEIJLHHSZRI-UHFFFAOYSA-N

storage temp.

Store at 2-8 C

Purity

98%

Shipping Temperature

RT

Physical Form

White crystalline powder

Product Link

https://www.chemimpex.com/product/productinfo/-i-n-i-1-ethoxyethylidene-2-aminooxyacetic-acid-i-n-i-hydroxysuccinimide-ester/23123?cid=1

Melting Point

53 - 62 C

MSDS

https://www.chemimpex.com/product/productinfo/-i-n-i-1-ethoxyethylidene-2-aminooxyacetic-acid-i-n-i-hydroxysuccinimide-ester/23123?cid=1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service