Skip to Content
Merck
All Photos(1)

Key Documents

AMBH97BA0994

Ambeed, Inc.

5-Bromo-3-methylfuran-2-carboxylic acid

Sign Into View Organizational & Contract Pricing

Select a Size

250 MG
US$17.25
1 G
US$50.60
5 G
US$241.50
25 G
US$846.40

Vendor SKU: A463932-250MG

US$17.25


Ships in 7 days - from Aldrich Partner.

To order products, please contact your local dealer.

Select a Size

Change View
250 MG
US$17.25
1 G
US$50.60
5 G
US$241.50
25 G
US$846.40

About This Item

CAS Number:
Molecular Weight:
205.01
MDL number:

Vendor SKU: A463932-250MG

US$17.25


Ships in 7 days - from Aldrich Partner.

To order products, please contact your local dealer.

Iupac Name

5-bromo-3-methyl-2-furoic acid

Inchi Code

1S/C6H5BrO3/c1-3-2-4(7)10-5(3)6(8)9/h2H,1H3,(H,8,9)

InChI key

WXEJPQJVASKGHW-UHFFFAOYSA-N

storage temp.

Inert atmosphere,2-8C

Purity

98%

Country of Origin

CN

Shipping Temperature

Normal

Physical Form

solid

Product Link

https://www.ambeed.com/products/5896-35-5.html

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service