Skip to Content
Merck
All Photos(1)

Key Documents

809446

Sigma-Aldrich

Poly(2-ethyl-2-oxazoline), α-benzyl, ω-thiol terminated

average Mn 10,000, PDI ≤1.3

Synonym(s):

PEtOx, POx, Polyoxazoline

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH2(C5H9NO)nSH
UNSPSC Code:
12352200
Pricing and availability is not currently available.

form

powder

mol wt

average Mn 10,000

PDI

≤1.3

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

Application

Poly(2-ethyl-2-oxazoline) is a hydrophilic polymer that is commonly used in biomedical applications.[1] This is a biocompatible polymer with one chain-end functionalized with a thiol group which is commonly used in ligation as well as in thiol-ene click chemistry. These polymers are commonly used as an amorphous alternative to PEGs.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Poly(2-oxazoline)s POx in Biomedical Applications.
Luxenhofer R and Jordan R
Material Matters, 8(3), 70-72 null

Articles

PiPrOx-based polymers exhibit diverse stimuli-responsive properties, showcased in recent developments for functional polymer systems.

The introduction of polymers into the biomedical field has opened new avenues in tissue engineering, implant design, biosensing, and drug delivery.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service