Skip to Content
Merck
All Photos(1)

Key Documents

771589

Sigma-Aldrich

Bis[2-(2-pyridinyl-N)phenyl-C](2,4-pentanedionato-O2,O4)iridium(III)

≥99.0% (HPLC)

Synonym(s):

(ppy)2Ir(acac), Bis[2-(2-pyridinyl-N)phenyl-C](acetylacetonato)iridium(III), Ir(ppy)2(acac)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H23IrN2O2
CAS Number:
Molecular Weight:
599.70
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

Pricing and availability is not currently available.

Assay

≥99.0% (HPLC)

form

powder

mp

349-356 °C

λmax

261 nm in dichloromethane

fluorescence

λem 314 nm in THF

SMILES string

CC(=O)[CH-]C(C)=O.[Ir+](c1ccccc1-c2ccccn2)c3ccccc3-c4ccccn4

InChI

1S/2C11H8N.C5H7O2.Ir/c2*1-2-6-10(7-3-1)11-8-4-5-9-12-11;1-4(6)3-5(2)7;/h2*1-6,8-9H;3H,1-2H3;/q;;-1;+1

InChI key

MCAGFPRRYUPGMD-UHFFFAOYSA-N

Application

(ppy)2Ir(acac) used in green phosphorescent OLED devices as dopant/emitter achieved high brightness of 130800 cd m_2 at a current density of 1550 mA cm_2 (13 V), and high quantum efficiency (15~16%).

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Phenylcarbazole-dipyridyl triazole hybrid as bipolar host material for phosphorescent OLEDs"
Journal of Materials Chemistry, 22, 5410-5418 (2012)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service