Skip to Content
Merck
All Photos(1)

Key Documents

768812

Sigma-Aldrich

1-Hydroxy-1,2-benziodoxole-3(1H)-one 1-oxide compound with pyridine (1:1)

≥90.0% (HPLC)

Synonym(s):

IBX pyridine complex, PIBX, Pyridinium o-iodoxybenzoate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H10INO4
CAS Number:
Molecular Weight:
359.12
Beilstein:
22776291
UNSPSC Code:
12352101
PubChem Substance ID:
NACRES:
NA.22
form:
powder
Assay:
≥90.0% (HPLC)
Pricing and availability is not currently available.

Assay

≥90.0% (HPLC)

form

powder

functional group

iodo

SMILES string

O=C(O1)C2=CC=CC=C2I1([O-])=O.C3=C[NH+]=CC=C3

InChI

1S/C7H5IO4.C5H5N/c9-7-5-3-1-2-4-6(5)8(10,11)12-7;1-2-4-6-5-3-1/h1-4H,(H,10,11);1-5H

InChI key

JKZNBOGIALICGN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Supplementary Hazards

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Pyridinium o-iodoxybenzoate as a safe form of a famous oxidant.
Kumanyaev, I. M., et al.
Mendeleev Communications, 22, 129-131 (2012)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service