Skip to Content
Merck
All Photos(1)

Documents

675539

Sigma-Aldrich

1,2-Bis[(2S,5S)-2,5-dimethylphospholano]benzene(cyclooctadiene)rhodium(I) tetrafluoroborate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C26H40BF4P2Rh
CAS Number:
Molecular Weight:
604.25
UNSPSC Code:
12352005
PubChem Substance ID:

form

solid

SMILES string

[F-].FB(F)F.C1CC=CCCC=C1.C[C@H]2CC[C@H](C)[PH]23c4ccccc4[PH]5([C@@H](C)CC[C@@H]5C)[Rh+]3(C)C

InChI

1S/C18H28P2.C8H12.2CH3.BF3.FH.Rh/c1-13-9-10-14(2)19(13)17-7-5-6-8-18(17)20-15(3)11-12-16(20)4;1-2-4-6-8-7-5-3-1;;;2-1(3)4;;/h5-8,13-16H,9-12H2,1-4H3;1-2,7-8H,3-6H2;2*1H3;;1H;/q;;;;;;-1/p+1/b;2-1-,8-7-;;;;;/t13-,14-,15-,16-;;;;;;/m0....../s1

InChI key

TVGPORVCUKHBTJ-SKAPDIIBSA-O

Legal Information

Sold in collaboration with Kanata Chemical Technologies Inc. for research purposes only. These compounds were made and sold under license from E.I. du Pont de Nemours and Company, which license does not include the right to use the compounds in producing products for sale in the pharmaceutical field.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Asymmetric hydrogenation enables scalable synthesis of single-enantiomer compounds with minimal byproducts, ideal for commercial manufacturing.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service