Skip to Content
Merck
All Photos(1)

Documents

465658

Sigma-Aldrich

N-(tert-Butoxycarbonyl)-L-threonine methyl ester

95%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH(OH)CH[NHCO2C(CH3)3]CO2CH3
CAS Number:
Molecular Weight:
233.26
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.22

Assay

95%

reaction suitability

reaction type: solution phase peptide synthesis

refractive index

n20/D 1.45 (lit.)

bp

206 °C (lit.)

application(s)

peptide synthesis

SMILES string

COC(=O)[C@@H](NC(=O)OC(C)(C)C)[C@@H](C)O

InChI

1S/C10H19NO5/c1-6(12)7(8(13)15-5)11-9(14)16-10(2,3)4/h6-7,12H,1-5H3,(H,11,14)/t6-,7+/m1/s1

InChI key

MZMWAPNVRMDIPS-RQJHMYQMSA-N

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service