Skip to Content
Merck
All Photos(1)

Documents

444103

Sigma-Aldrich

Dysprosium(III) perchlorate solution

40 wt. % in H2O, 99.9% trace metals basis

Synonym(s):

Dysprosium perchlorate (Dy(ClO4 )3 ), Dysprosium triperchlorate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Dy(ClO4)3
CAS Number:
Molecular Weight:
460.85
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:

vapor pressure

0.12 psi ( 20 °C)
1.68 psi ( 55 °C)

Assay

99.9% trace metals basis

form

liquid

reaction suitability

reagent type: catalyst
core: dysprosium

concentration

40 wt. % in H2O

density

1.476 g/mL at 25 °C

SMILES string

[Dy+3].[O-]Cl(=O)(=O)=O.[O-]Cl(=O)(=O)=O.[O-]Cl(=O)(=O)=O

InChI

1S/3ClHO4.Dy/c3*2-1(3,4)5;/h3*(H,2,3,4,5);/q;;;+3/p-3

InChI key

QNDHOIIJUAAORW-UHFFFAOYSA-K


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service