Skip to Content
Merck
All Photos(1)

Documents

436992

Sigma-Aldrich

N-Boc-1,3-propanediamine

95%

Synonym(s):

N-Boc-1,3-diaminopropane, tert-Butyl N-(3-aminopropyl)carbamate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)3COCONH(CH2)3NH2
CAS Number:
Molecular Weight:
174.24
Beilstein:
3588328
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

Assay

95%

reaction suitability

reagent type: cross-linking reagent
reagent type: spacer

refractive index

n20/D 1.454 (lit.)

bp

203 °C (lit.)

mp

22 °C (lit.)

density

0.998 g/mL at 20 °C (lit.)

SMILES string

NCCCNC(OC(C)(C)C)=O

InChI

1S/C8H18N2O2/c1-8(2,3)12-7(11)10-6-4-5-9/h4-6,9H2,1-3H3,(H,10,11)

InChI key

POHWAQLZBIMPRN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

replaced by

Product No.
Description
Pricing

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

228.2 °F - closed cup

Flash Point(C)

109 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service