295159
Chloropentafluoroethane
≥98%
Synonym(s):
CFC-115
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C2ClF5
CAS Number:
Molecular Weight:
154.47
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:
Recommended Products
Quality Level
Assay
≥98%
functional group
chloro
fluoro
SMILES string
FC(F)(F)C(F)(F)Cl
InChI
1S/C2ClF5/c3-1(4,5)2(6,7)8
InChI key
RFCAUADVODFSLZ-UHFFFAOYSA-N
Packaging
Packaged in steel cylinders
Recommended products
Brass regulator Z146706 or Z146714 is recommended.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
A C Fernandes et al.
Radiation protection dosimetry, 120(1-4), 503-508 (2006-04-29)
We describe the fabrication and characterisation of the SIMPLE superheated droplet detector, a 10 g active mass device of C(2)ClF(5) in 1-3% weight concentrations currently employed in a direct search for spin-dependent astroparticle dark matter candidates.
R L Fitzgerald et al.
Journal of forensic sciences, 38(2), 477-483 (1993-03-01)
Reports on fatalities of chlorofluorocarbons usually involve chlorotrifluoroethane, trichlorofluoromethane, dichlorodifluoromethane or chlorodifluoromethane, where analysis was done using packed column gas chromatography. In this case a death was caused by an azeotropic mixture of chlorodifluoromethane and chloropentafluoroethane, a combination that has
F d'Errico et al.
Physics in medicine and biology, 43(5), 1147-1158 (1998-06-12)
A position-sensitive detector chamber is introduced for the three-dimensional (3D) dosimetry of photon-emitting brachytherapy sources. The detector is based on an extremely fine suspension of monochloropentafluoroethane droplets emulsified in a gel. The droplets are highly superheated at room temperature and
M Das et al.
Applied radiation and isotopes : including data, instrumentation and methods for use in agriculture, industry and medicine, 53(4-5), 759-763 (2000-09-26)
The present work relates to a sensitive neutron dosimeter, a device for monitoring neutron dose in some accelerator and reactor sites. This device is capable of measuring a neutron dose as small as 0.1 microSv using superheated liquid as a
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service