Skip to Content
Merck
All Photos(1)

Documents

290114

Sigma-Aldrich

2-Hydroxy-4-methyl-3-nitropyridine

98%

Synonym(s):

4-Methyl-3-nitro-2-pyridone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H6N2O3
CAS Number:
Molecular Weight:
154.12
Beilstein:
139125
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

solid

mp

229-232 °C (lit.)

SMILES string

Cc1ccnc(O)c1[N+]([O-])=O

InChI

1S/C6H6N2O3/c1-4-2-3-7-6(9)5(4)8(10)11/h2-3H,1H3,(H,7,9)

InChI key

HZCWTTHQRMHIIE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

The conformational stability and the vibartional analysis of 2-hydroxy-4-methyl-3-nitropyridine was studied.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Balachandran V, et al.
Journal of Molecular Structure, 1013, 75-85 (2012)
Jonita Stankevičiūtė et al.
Scientific reports, 6, 39129-39129 (2016-12-17)
Pyridinols and pyridinamines are important intermediates with many applications in chemical industry. The pyridine derivatives are in great demand as synthons for pharmaceutical products. Moreover, pyridines are used either as biologically active substances or as building blocks for polymers with

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service