All Photos(1)
About This Item
Linear Formula:
(H2NCH2CH2NH2)2PtCl2
CAS Number:
Molecular Weight:
386.18
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
reaction suitability
reagent type: catalyst
core: platinum
SMILES string
Cl[Pt]Cl.NCCN.NCCN
InChI
1S/2C2H8N2.2ClH.Pt/c2*3-1-2-4;;;/h2*1-4H2;2*1H;/q;;;;+2/p-2
InChI key
MXLIXEZAMQLDMM-UHFFFAOYSA-L
Application
Reactant for:
- Electroreduction reactions
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service