More Cytokines Means Even More Discoveries
Multiplex with MILLIPLEX® Human Cytokine Panels A & B
Cytokines, chemokines, and growth factors are key mediators of immune system functions capable of signaling through autocrine, paracrine, and endocrine mechanisms. Their pleiotropic immunomodulatory properties allow these biomolecules to react to diverse stimuli and regulate the immune response, either promoting or inhibiting inflammation.
MILLIPLEX® Human Cytokine/Chemokine/Growth Factor Panel A and Panel B
The open-architecture xMAP® technology of the MILLIPLEX® Human Cytokine/Chemokine/Growth Factor Panel A and Panel B enables multiplexing of many types of bioassays reducing time, labor, and costs over traditional methods. The capability of adding multiple conjugated beads to each sample results in the ability to obtain multiple cytokine results from each sample.
MILLIPLEX®Human Cytokine Panel A and B offer you:
- The ability to select a 38-plex or 48-plex premixed kit
- The flexibility to choose any combination of analytes from our panel of 48 analytes to design a custom kit that better meets your needs
- A convenient “all-in-one” box format that provides all the necessary reagents you need to run your assay
- Small sample volume requirements (25 μL/well)
- Superior performance with rigorous assay development and quality control testing
Panel Details
These panels are 48-plex kits to be used for the simultaneous quantification of any or all of the following analytes in serum or plasma samples and tissue/cell lysate and culture supernatant samples:
- MILLIPLEX® Human Cytokine/Chemokine/Growth Factor Panel A: sCD40L, EGF, Eotaxin, FGF-2, FLT-3L, Fractalkine, G-CSF, GM-CSF, GROα, IFNα2, IFNγ, IL-1α, IL-1β, IL-1RA, IL-2, IL-3, IL-4, IL-5, IL-6, IL-7, IL-8, IL-9, IL-10, IL-12 (p40), IL-12 (p70), IL-13, IL-15, IL-17A, IL-17E/IL-25, IL-17F, IL-18, IL-22, IL-27, IP-10, MCP-1, MCP-3, M-CSF, MDC, MIG, MIP-1α, MIP-1β, PDGF-AA, PDGF-AB/BB, RANTES*, TGFα, TNFα, TNFβ, VEGF-A
- MILLIPLEX® Human Cytokine/Chemokine/Growth Factor Panel B: 6Ckine, sCD137, sFAS, sFASL, APRIL, BAFF, BCA-1, CCL28, CTACK, CXCL16, ENA-78, Eotaxin-2, Eotaxin-3, GCP-2, Granzyme A, Granzyme B, HMGB1, I-309, IFNβ, IFNω, IL-11, IL-16, IL-20, IL-21, IL-23, IL-24, IL-28A, IL-29, IL-31, IL-33, IL-34, IL-35, I-TAC, LIF, Lymphotactin, MCP-2, MCP-4, MIP-1δ, MIP-3α, MIP-3β, MPIF-1, Perforin, SCF, SDF-1, TARC, TPO, TRAIL, TSLP
- MILLIPLEX® Human Cytokine Panel A + B Combo Packs: Choose a 76-plex combo pack containing the Panel A 38-plex premixed kit and the Panel B 38-plex premixed kit or a 96-plex combo pack containing the Panel A 48-plex premixed kit and the Panel B 48-plex premixed kit. Combo packs offer convenient ordering and save you money!
* RANTES is always provided as a separate bead vial due to different dilution requirements for serum/ plasma samples. If measuring RANTES in serum/plasma, it is recommended to use a singleplex kit including RANTES only.
Network error: Failed to fetch
Related Resources
- Next-Gen Cytokine Multiplex Assays
MILLIPLEX® cytokine multiplex assays enable comprehensive immune system investigation.
- Cytokine Measurement Consistency
MILLIPLEX® multiplex cytokine assay demonstrates superior lot-to-lot consistency in immunoassays for reliable results.
- Immunology Multiplex Assays | MILLIPLEX®
Immunology multiplex assays like MILLIPLEX® allow simultaneous investigation of immune biomarkers, saving time and resources in research.
- Neuroimmunology & Neuroinflammation Immunoassays
Find immunoassays to explore neuroimmunology and neuroinflammation cytokines and neurodegenerative disease biomarkers.
- Quality in MILLIPLEX® Multiplex Kits
Learn more about how you can rely on the quality we build into each MILLIPLEX® multiplex kit to produce results you trust.
- Overview of Luminex® Multiplex Assay Technology | MILLIPLEX® Multiplex Assays
Discover the benefits of MILLIPLEX® multiplex assays, based on Luminex® xMAP® multiplex assay technology, that provide consistent, high-quality results and see how these multiplex biomarker Luminex® assays are being used to advance research.