Merck
Find Simplicity in Stability
Streamline Your CHO Genetic Stability Testing with the Aptegra™ Platform
Aptegra™ CHO Genetic Stability Assay
Streamline Your Genetic Stability Testing with the NGS-based Aptegra™ Platform
The genetic stability testing package for biopharmaceutical cell banks has remained relatively unchanged for decades. This traditional approach to genetic stability testing relies on a complex set of assays to meet ICH Q5B requirements, many of which have limitations in discovery power and sensitivity and require long timelines for completion. 
The proprietary Aptegra™ platform delivers the first streamlined, single-test replacement of traditional methods with the Aptegra™ CHO genetic stability assay delivering results in a third of the time. This assay taps into the combined power of whole genome sequencing (WGS) and expert bioinformatics to deliver a faster, all-in-one solution for a comprehensive testing package, including: 
  • Gene of interest (GOI) sequence analysis
  • Copy number determination
  • Sequence analysis of flanking regions
  • Integration site determination
  • Analysis of insertions or deletions
The Aptegra™ CHO genetic stability assay operates under a GMP-validated workflow to return expert-reviewed results in 30 days using less of your precious cell bank. 
Stability testing should be simple, by tapping into the whole genome, it can be. The Aptegra™ CHO genetic stability assay:
  • Replaces Southern, Sanger sequencing, PCR, and FISH with a single technology
  • Utilizes expert bioinformatics to address all genetic stability regulatory requirements
  • Eliminates primer bias, design time, and assay optimization
  • Eradicates any guesswork in data interpretation
  • Reduces upfront client time with off-the-shelf ordering
  • Consistently delivers fast, reliable results
Aptegra™ CHO Genetic Stability assay vs. the traditional package of five assays
Aptegra™ CHO Genetic Stability assay
Comprehensive Traditional Package
Number of assays recommended​
1
​5
Multiple genes tested per assay​
Yes
No​
Proposal required​
No
Yes​
Development work / primer design​
No
Yes​
Client time investment ​
Sample Submission
Project kick-off meetings as needed​
Sample requirement ​
2.0 x 106 cells
2.3 x 108 cells​
Growing cells required 
No
Yes*​
Side-by-side sample testing for EOPC​
No
Yes**​
Time to complete project​
30 days​
3-5 months (minimum)​
* Actively growing cells required for FISH analysis​
** Recommended for ddPCR and Southern blot assays
Simplify your genetic stability testing with the Aptegra™ platform.
Talk to an expert today