Merck
HomePolymerase Chain Reaction ApplicationsDirect PCR — Extract-N-Amp™ PCR Master Mix
Direct PCR — Extract-N-Amp™ PCR Master Mix
What is direct PCR?
Direct PCR is a method to directly amplify DNA from plant, blood, and tissue without the need for traditional DNA extraction and DNA purification steps. With the removal of often laborious and time-consuming DNA isolation steps, direct PCR kits enable scientists to save both resources in the lab and time at the bench. Explore the features and benefits of our direct PCR kits, Extract-N-Amp™ PCR master mix reagents, and discover additional resources and protocols for your plant, blood, and tissue DNA amplification workflow needs.
Extract-N-Amp™ Direct PCR Product Guide
Leveraging the robust benefits of hot start PCR technology, Extract-N-Amp™ PCR kits are the world’s first integrated extraction and amplification process for rapid blood, tissue, and plant assays. Eliminate the need for columns or long enzymatic sample purification with this simple “lyse & go” method for PCR. Our kits include all the reagents, proprietary buffers, and enzymes needed to rapidly extract and amplify targets of interest from a variety of cells and tissues.
REDExtract-N-Amp™ Direct PCR Workflow Video
Extract-N-Amp™ PCR technology eliminates the need for long enzymatic digestions or homogenization by incorporating a novel extraction method. The kits also include a specially formulated hot start PCR ReadyMix for amplification directly from the extract. The PCR ReadyMix comes in two formulations: Extract-N-Amp™ PCR ReadyMix and REDExtract-N-Amp™ PCR ReadyMix.
REDExtract-N-Amp™ Direct PCR ReadyMix
The REDExtract-N-Amp™ PCR ReadyMix contains an inert dye that acts as a tracking dye and allows for convenient loading of PCR reaction onto agarose gels for analysis. REDExtract-N-Amp™ PCR ReadyMix is suitable for multiple routine applications, including genotyping, transgene detection, knockout analysis, and sequencing. In a typical procedure, genomic DNA is extracted from a sample that has been incubated in preparation solution and extraction solution at room temperature. The sample is heated and then mixed with a third solution to neutralize inhibitory substances prior to PCR. A portion of the DNA extract is then added to a PCR reaction containing primers and the ReadyMix included in the kit. The remainder of the DNA can be refrigerated for convenient storage.
Extract-N-Amp™ Direct PCR Protocols, PCR Troubleshooting, and Resources
Explore our validated protocols and resources to extract and amplify DNA from tissue, blood, and plant samples. Additionally, our convenient PCR master mix, buffers, and recipe calculators resource hub offers tools for your PCR workflow needs.
ProductSpeedProduct DescriptionStarting Material
Extract-N-Amp™ Tissue PCR Kit
Extract-N-Amp™ REDExtract-N-Amp Tissue PCR Kit
Less than 15 minutes from extraction to amplification
Rapid amplification of genomic DNA from
  • Mouse-tails
  • Hair
  • Animal Tissue
  • Buccal Swabs
Tail: 0.5 to 1.0 cm
Hair: 1 root
Tissue: 2 to 10 mg
Buccal: 10 µL
SYBR® Green Extract-N-Amp™ Tissue PCR KitLess than 15 minutes from extraction to amplification
Rapid quantitative PCR of genomic DNA from
  • Mouse-tails
  • Hair
  • Animal Tissue
  • Buccal Swabs
Tail: 0.5 to 1.0 cm
Hair: 1 root
Tissue: 2 to 10 mg
Buccal: 10 µL
Extract-N-Amp™ Blood PCR Kit
REDExtract-N-Amp Blood PCR Kit
8 minutes from extraction to amplificationRapid amplification of genomic DNA from
  • Whole blood
Blood: 10 µL
Extract-N-Amp™ Plant PCR Kit Protocol
Extract-N-Amp™ Plant Tissue Sample Data
REDExtract-N-Amp Plant PCR Kit
Less than 15 minutes from extraction to amplificationRapid extraction and PCR amplification of genomic DNA from
  • Plant leaves
Plant Leaf Disk:
0.5 to 0.7 cm
SYBR® Green Extract-N-Amp™ Plant PCR KitLess than 15 minutes from extraction to amplificationRapid extraction and PCR amplification of genomic DNA from
  • Plant leaves
Plant Leaf Disk:
0.5 to 0.7 cm
Extract-N-Amp™ Seed PCR Kit
REDExtract-N-Amp Seed PCR Kit
Less than 15 minutes from extraction to amplificationRapid amplification of genomic DNA from
  • Plant seeds
1 seed
Materials
Sorry, an unexpected error has occurred
Network error: Failed to fetch