- Development and characterization of an α-l-rhamnosidase mutant with improved thermostability and a higher efficiency for debittering orange juice.
Development and characterization of an α-l-rhamnosidase mutant with improved thermostability and a higher efficiency for debittering orange juice.
The glycoside hydrolase, α-l-rhamnosidase, could remove the bitter taste of naringin from citrus juices. However, most α-l-rhamnosidases are easily deactivated at high temperatures, limiting the practice in debittering citrus juices. The V529A mutant of the α-l-rhamnosidase r-Rha1 from Aspergillus niger JMU-TS528 was developed with improved thermostability using directed evolution technology and site-directed mutagenesis. The enzyme mutant had a half-live of thermal inactivation T