Merck
  • Secreted CLIC3 drives cancer progression through its glutathione-dependent oxidoreductase activity.
Secreted CLIC3 drives cancer progression through its glutathione-dependent oxidoreductase activity.
Nature communications (2017-02-16)
Juan R Hernandez-Fernaud, Elena Ruengeler, Andrea Casazza, Lisa J Neilson, Ellie Pulleine, Alice Santi, Shehab Ismail, Sergio Lilla, Sandeep Dhayade, Iain R MacPherson, Iain McNeish, Darren Ennis, Hala Ali, Fernanda G Kugeratski, Heba Al Khamici, Maartje van den Biggelaar, Peter V E van den Berghe, Catherine Cloix, Laura McDonald, David Millan, Aoisha Hoyle, Anna Kuchnio, Peter Carmeliet, Stella M Valenzuela, Karen Blyth, Huabing Yin, Massimiliano Mazzone, Jim C Norman, Sara Zanivan
ABSTRACT
The secretome of cancer and stromal cells generates a microenvironment that contributes to tumour cell invasion and angiogenesis. Here we compare the secretome of human mammary normal and cancer-associated fibroblasts (CAFs). We discover that the chloride intracellular channel protein 3 (CLIC3) is an abundant component of the CAF secretome. Secreted CLIC3 promotes invasive behaviour of endothelial cells to drive angiogenesis and increases invasiveness of cancer cells both in vivo and in 3D cell culture models, and this requires active transglutaminase-2 (TGM2). CLIC3 acts as a glutathione-dependent oxidoreductase that reduces TGM2 and regulates TGM2 binding to its cofactors. Finally, CLIC3 is also secreted by cancer cells, is abundant in the stromal and tumour compartments of aggressive ovarian cancers and its levels correlate with poor clinical outcome. This work reveals a previously undescribed invasive mechanism whereby the secretion of a glutathione-dependent oxidoreductase drives angiogenesis and cancer progression by promoting TGM2-dependent invasion.
Sigma-Aldrich
Anti-TGM2 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Benzamidine, ≥95.0%
Sigma-Aldrich
Monoclonal Anti-Vinculin antibody produced in mouse, clone hVIN-1, ascites fluid
Sigma-Aldrich
Anti-Actin, α-Smooth Muscle antibody, Mouse monoclonal, clone 1A4, purified from hybridoma cell culture
Sigma-Aldrich
Anti-Laminin-5 (γ2 chain) Antibody, clone D4B5, clone D4B5, Chemicon®, from mouse