Merck
  • Targeting Wnt/β-Catenin Activated Cells with Dominant-Negative N-cadherin to Reduce Neointima Formation.
Targeting Wnt/β-Catenin Activated Cells with Dominant-Negative N-cadherin to Reduce Neointima Formation.
Molecular therapy. Methods & clinical development (2017-05-26)
Sarah Hulin-Curtis, Helen Williams, Kerry S Wadey, Graciela B Sala-Newby, Sarah J George
ABSTRACT
Approximately 50% of coronary artery bypass grafts using the autologous saphenous vein fail within 10 years due to intimal thickening. This study examined whether a gene therapy approach that selectively kills Wnt/β-catenin/T cell factor (TCF) activated vascular smooth muscle cells (VSMCs) using dominant-negative N-cadherin (dn-N-cadherin) reduced intimal thickening. Cultured human VSMCs infected with an adenovirus (Ad) encoding dn-N-cadherin via the TCF promoter (Ad-TOP-dn-N-cadherin) specifically expressed dn-N-cadherin in response to activation of the Wnt/β-catenin/TCF pathway. Infection with Ad-TOP-dn-N-cadherin significantly increased VSMC apoptosis (3 ± 0.2% versus 9 ± 0.7%; p < 0.05, n = 6) and significantly inhibited VSMC migration by 83 ± 15% (p < 0.05, n = 6), but did not affect VSMC proliferation (p > 0.05, n = 5). In an ex vivo human saphenous vein organ culture model, luminal delivery of Ad-TOP-dn-N-cadherin significantly increased VSMC apoptosis after 7 days of culture (4 ± 1.4% versus 9 ± 1.6%; p < 0.01, n = 6) and suppressed intimal thickening by 75 ± 7% (p < 0.05, n = 5), without a detrimental effect on endothelial cell coverage. In vivo, Ad-TOP-dn-N-cadherin significantly reduced intimal thickening at day 21 (n = 10) in comparison to the Ad-β-galactosidase (Ad-β-gal) control virus (n = 12, p < 0.05) in the mouse carotid artery ligation model. In summary, we have developed a novel approach to selectively reduce intimal thickening, which may be beneficial in reducing late vein graft failure.
Sigma-Aldrich
Monoclonal Anti-BrdU antibody produced in mouse, clone BU-33, ascites fluid, Immunohistology Grade
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-74, ascites fluid