Merck
  • The developmental origin of zygodactyl feet and its possible loss in the evolution of Passeriformes.
The developmental origin of zygodactyl feet and its possible loss in the evolution of Passeriformes.
Proceedings. Biological sciences (2014-06-27)
João Francisco Botelho, Daniel Smith-Paredes, Daniel Nuñez-Leon, Sergio Soto-Acuña, Alexander O Vargas
ABSTRACT
The zygodactyl orientation of toes (digits II and III pointing forwards, digits I and IV pointing backwards) evolved independently in different extant bird taxa. To understand the origin of this trait in modern birds, we investigated the development of the zygodactyl foot of the budgerigar (Psittaciformes). We compared its muscular development with that of the anisodactyl quail (Galliformes) and show that while the musculus abductor digiti IV (ABDIV) becomes strongly developed at HH36 in both species, the musculus extensor brevis digiti IV (EBDIV) degenerates and almost disappears only in the budgerigar. The asymmetric action of those muscles early in the development of the budgerigar foot causes retroversion of digit IV (dIV). Paralysed budgerigar embryos do not revert dIV and are anisodactyl. Both molecular phylogenetic analysis and palaeontological information suggest that the ancestor of passerines could have been zygodactyl. We followed the development of the zebra finch (Passeriformes) foot muscles and found that in this species, both the primordia of the ABDIV and of the EBDIV fail to develop. These data suggest that loss of asymmetric forces of muscular activity exerted on dIV, caused by the absence of the ABDIV, could have resulted in secondary anisodactyly in Passeriformes.
Product Description
Roche
DAB Substrate, dark brown, /black precipitate; Visually evaluated
Pricing and availability is not currently available.