Merck
  • Inhibition of p53 increases chemosensitivity to 5-FU in nutrient-deprived hepatocarcinoma cells by suppressing autophagy.
Inhibition of p53 increases chemosensitivity to 5-FU in nutrient-deprived hepatocarcinoma cells by suppressing autophagy.
Cancer letters (2014-01-28)
Xian-ling Guo, Fei Hu, Shan-shan Zhang, Qiu-dong Zhao, Chen Zong, Fei Ye, Shi-wei Guo, Jian-wei Zhang, Rong Li, Meng-chao Wu, Li-xin Wei
ABSTRACT
Activation of p53 can induce apoptosis, cell cycle arrest, and cell senescence, although some evidence has suggested that p53 could promote cell survival. However, whether p53 plays a positive role in cancer cell survival to chemotherapy remains unknown. In this study, we show that inhibition of p53 enhanced apoptosis and increased chemosensitivity to 5-fluorouracil (5-FU) in nutrient-deprived hepatocarcinoma cells (HCC). Meanwhile, nutrient-deprivation-induced autophagy was inhibited by pifithrin-α or small interfering RNA targeting p53. The expression of p53 was not increased when HCC were incubated under nutrient-deprived conditions. This indicates that the basal level of p53 is important to autophagy activation in nutrient-deprived HCC cells. Furthermore, combining p53 inhibition and nutrient deprivation or 5-FU treatment resulted in a marked increase in reactive oxygen species generation and mitochondrial damage. Antioxidants reduced nutrient deprivation or 5-FU-induced cell death of HCC after p53 inhibition. Our results suggest that p53 contributes to cell survival and chemoresistance in HCC under nutrient-deprived conditions by modulating autophagy activation.
Product Description
Supelco
Toluene solution, certified reference material, 5000 μg/mL in methanol
Supelco
Toluene solution, certified reference material, TraceCERT®, 200 μg/mL in methanol, ampule of 1 mL
Sigma-Aldrich
Toluene, suitable for HPLC, ≥99.9%
Supelco
Residual Solvent - Toluene, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Toluene, analytical standard
Sigma-Aldrich
Toluene, anhydrous, 99.8%
Supelco
Toluene, HPLC grade, 99.8%
Sigma-Aldrich
Toluene, JIS 1000, for residue analysis, ≥99.8%
Sigma-Aldrich
Toluene, JIS 300, for residue analysis, ≥99.8%
Sigma-Aldrich
Toluene, JIS special grade, ≥99.5%
Sigma-Aldrich
Toluene, SAJ first grade, ≥99.0%
Sigma-Aldrich
Toluene, ACS reagent, ≥99.5%
Sigma-Aldrich
Toluene, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.7% (GC)
Sigma-Aldrich
Toluene, ACS reagent, ≥99.5%
Sigma-Aldrich
Toluene, ACS reagent, ≥99.5%
Sigma-Aldrich
Toluene, Laboratory Reagent, ≥99.3%
Supelco
Toluene, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Toluene, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%
Sigma-Aldrich
Toluene, suitable for HPLC, 99.9%