Merck
  • Heparanase-mediated cleavage of macromolecular heparin accelerates release of granular components of mast cells from extracellular matrices.
Heparanase-mediated cleavage of macromolecular heparin accelerates release of granular components of mast cells from extracellular matrices.
The Biochemical journal (2013-12-19)
Nobuaki Higashi, Michihiko Waki, Mayumi Sue, Yusuke Kogane, Hiroaki Shida, Naoki Tsunekawa, Ahasanul Hasan, Takeshi Sato, Ayumi Kitahara, Tatsuhiko Kasaoka, Yoshihiro Hayakawa, Motowo Nakajima, Tatsuro Irimura
ABSTRACT
Heparanase cleaves macromolecular heparin in the secretory granules of connective tissue-type mast cells. We investigated roles of the cleavage under a microenvironment mimicking where the mast cells physiologically reside. A connective tissue-type mast cell line MST and mouse peritoneal cell-derived mast cells stored macromolecular heparin in the secretory granules. The cells expressing heparanase stored fragmented heparin (~10 kDa) due to heparanase-dependent cleavage of the heparin. We produced an artificial collagen-based extracellular matrix and placed the live cells or glycosaminoglycans purified from the cells in the matrix to measure the release of sulfated macromolecules into the medium. The sulfate-radiolabelled molecules from the degranulating heparanase-expressing cells and the purified glycosaminoglycans showed significantly greater release into the medium than those derived from mock cells, which was not the case in suspension culture. The mast cell granular enzyme chymase, but not β-hexosaminidase, showed significantly greater release from the degranulating heparanase-expressing cells than from mock cells. Purified chymase mixed with fragmented heparin derived from heparanase-expressing cells showed greater release from collagen gels than the enzyme alone or mixed with macromolecular heparin derived from mock cells. We propose that the cleavage of macromolecular heparin by heparanase accelerates the release of heparin and chymase from extracellular matrices.
Sigma-Aldrich
Chymase human, recombinant, expressed in Pichia pastoris
Sigma-Aldrich
β-Glucuronidase from Helix pomatia, Type H-3, aqueous solution, ≥90,000 units/mL
Sigma-Aldrich
β-Glucuronidase from Helix pomatia, Type H-3AF, aqueous solution, ≥60,000 units/mL
Sigma-Aldrich
β-Glucuronidase from limpets (Patella vulgata), Type L-II, lyophilized powder, 1,000,000-3,000,000 units/g solid
Sigma-Aldrich
β-Glucuronidase from Helix pomatia, Type HP-2, aqueous solution, ≥100,000 units/mL
Sigma-Aldrich
β-Glucuronidase from Helix pomatia, Type H-2, aqueous solution, ≥85,000 units/mL
Sigma-Aldrich
β-Glucuronidase from Helix pomatia, Type H-1, partially purified powder, ≥300,000 units/g solid
Sigma-Aldrich
β-Glucuronidase from Helix pomatia, Type HP-2S, aqueous solution, ≥90,000 units/mL
Sigma-Aldrich
β-Glucuronidase from Helix pomatia, Type H-5, lyophilized powder, ≥400,000 units/g solid
Sigma-Aldrich
β-Glucuronidase from limpets (Patella vulgata), aqueous solution, ≥85,000 units/mL
Sigma-Aldrich
β-Glucuronidase from Escherichia coli, ≥20,000 units/mg protein, recombinant, expressed in E. coli overproducing strain, lyophilized powder
Sigma-Aldrich
β-Glucuronidase from Helix aspersa (garden snail), Type HA-4
Sigma-Aldrich
β-Glucuronidase from bovine liver, Type B-3, ≥2,000,000 units/g solid
Sigma-Aldrich
β-Glucuronidase from Escherichia coli, Type IX-A, lyophilized powder, 1,000,000-5,000,000 units/g protein (30 min assay)
Sigma-Aldrich
β-Glucuronidase from Escherichia coli, aqueous glycerol solution, ≥5,000,000 units/g protein, pH 6.8 (biuret)
Sigma-Aldrich
β-Glucuronidase from bovine liver, Type B-1, ≥1,000,000 units/g solid
Sigma-Aldrich
β-Glucuronidase from Escherichia coli, Type VII-A, lyophilized powder, 5,000,000-20,000,000 units/g protein, pH 6.8 (30 min assay)
Sigma-Aldrich
β-Glucuronidase from Escherichia coli, >20,000,000 units/g protein, recombinant, expressed in E. coli, aqueous glycerol solution
Sigma-Aldrich
β-Glucuronidase from Escherichia coli, ≥10,000,000 units/g protein (30 min assay), recombinant, expressed in E. coli overproducing strain, lyophilized powder