- Methyl 3-(4-methyl-benzyl-idene)carbazate.
Methyl 3-(4-methyl-benzyl-idene)carbazate.
Acta crystallographica. Section E, Structure reports online (2010-01-01)
Yu-Feng Li, Fu-Gong Zhang, Fang-Fang Jian
PMID21579505
ABSTRACT
The title compound, C(10)H(12)N(2)O(2), was prepared by the reaction of methyl carbazate and 4-methyl-benzaldehyde. The dihedral angle between the benzene ring and the carbazate fragment is 20.86 (10)°. In the crystal structure, mol-ecules are linked by inter-molecular N-H⋯O hydrogen bonds.