Merck
  • In vitro transformation of hamster embryo cells by 3-(N-salicyloyl)amino-1,2,4-triazole.
In vitro transformation of hamster embryo cells by 3-(N-salicyloyl)amino-1,2,4-triazole.
Toxicology letters (1981-01-01)
K Inoue, Y Katoh, S Takayama
ABSTRACT
The in vitro carcinogenic activities of 3-(N-salicyloyl)amino-1,2,4-triazole (SAT) and its two components, 3-amino-1,2,4-triazole and phenyl salicylate were examined in a transformation assay with cryopreserved hamster embryo cells. SAT induced morphological transformation at certain doses between 10 microgram/ml and 100 microgram/ml in each of 5 repeated experiments, but gave a negative result in the Salmonella mutation assay. 3-Amino-1,2,4-triazole also induced transformation in 3 repeated experiments. Phenyl salicylate did not induce transformation at any of the doses tested in 3 consecutive experiments.
Product Description
Supelco
Phenyl salicylate melting point standard, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Phenyl salicylate, ReagentPlus®, 99%
Sigma-Aldrich
Phenyl salicylate, ≥99%, FG
Supelco
Mettler-Toledo Calibration substance ME 30034252, Phenyl salicylate, traceable to primary standards