Merck
  • Fungal volatiles associated with moldy grain in ventilated and non-ventilated bin-stored wheat.
Fungal volatiles associated with moldy grain in ventilated and non-ventilated bin-stored wheat.
Mycopathologia (1988-01-01)
R N Sinha, D Tuma, D Abramson, W E Muir
ABSTRACT
The fungal odor compounds 3-methyl-1-butanol, 1-octen-3-ol and 3-octanone were monitored in nine experimental bins in Winnipeg, Manitoba containing a hard red spring wheat during the autumn, winter and summer seasons of 1984-85. Quality changes were associated with seed-borne microflora and moisture content in both ventilated and non-ventilated bins containing wheat of 15.6 and 18.2% initial moisture content. All three odor compounds occurred in considerably greater amounts in bulk wheat in non-ventilated than in ventilated bins, particularly in those with wheat having 18.2% moisture content. The presence of these compounds usually coincided with infection of the seeds by the fungi Alternaria alternata (Fr.) Keissler, Aspergillus repens DeBarry, A. versicolor (Vuill.) Tiraboschi, Penicillium crustosum Thom, P. oxalicum Currie and Thom, P. aurantiogriesum Dierckx, and P. citrinum Thom. High production of all three odor compounds in damp wheat stored in non-ventilated bins was associated with heavy fungal infection of the seeds and reduction in seed germinability. High initial moisture content of the harvested grain accelerated the production of all three fungal volatiles in non-ventilated bins.
Product Description
Supelco
3-Octanone, analytical standard
Sigma-Aldrich
3-Octanone, ≥98%
Sigma-Aldrich
3-Octanone, ≥98%, FG