Merck
  • Phosphorescent imaging of living cells using a cyclometalated iridium (III) complex.
Phosphorescent imaging of living cells using a cyclometalated iridium (III) complex.
PloS one (2013-03-05)
Dik-Lung Ma, Hai-Jing Zhong, Wai-Chung Fu, Daniel Shiu-Hin Chan, Hiu-Yee Kwan, Wang-Fun Fong, Lai-Hon Chung, Chun-Yuen Wong, Chung-Hang Leung
ABSTRACT
A cell permeable cyclometalated iridium(III) complex has been developed as a phosphorescent probe for cell imaging. The iridium(III) solvato complex [Ir(phq)2(H2O]2)] preferentially stains the cytoplasm of both live and dead cells with a bright luminescence.
Sigma-Aldrich
Iridium, evaporation slug, diam. × L 0.6 cm × 1.2 cm, 99.9% trace metals basis
Sigma-Aldrich
Iridium, powder, 99.9% trace metals basis
Sigma-Aldrich
Iridium, foil, thickness 0.25 mm, 99.9% trace metals basis