Merck
  • Selective spectrofluorimetric determination of sulfide ion using manganese doped ZnS quantum dots as luminescent probe.
Selective spectrofluorimetric determination of sulfide ion using manganese doped ZnS quantum dots as luminescent probe.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy (2013-02-26)
Hamid Reza Rajabi, Mojtaba Shamsipur, Ali Azam Khosravi, Omid Khani, Mohammad Hasan Yousefi
ABSTRACT
This work reports a spectrofluorimetric method for selective and sensitive determination of sulfide ion in aqueous solution. The ultra-small zinc sulfide quantum dots (QDs) doped with manganese (ZnS:Mn) were synthesized by using a simple and fast procedure based on the co-precipitation of nanoparticles in aqueous solution in the presence of 2-mercaptoethanol, as capping agent. The nanoparticles have exhibited two strong fluorescent emissions at about 424 and 594 nm. Luminescent surface-capped ZnS:Mn QDs, with particle size below 5 nm, have been applied for determination of sulfide anions in water samples. Under the optimum conditions, the fluorescence intensity of ZnS:Mn QDs is linearly proportional to the sulfide ion concentration in the range 1.2×10(-6) to 2.6×10(-5) mol L(-1) with a detection limit as 3.3×10(-7) mol L(-1). The relative standard deviation for five replicate measurements (for 8.0×10(-6) mol L(-1) of S(2-)) was obtained to be 2.6%. It was founded that the interference of the other anions was negligible on the quantitive determination of sulfide ion.
Sigma-Aldrich
Zinc sulfide, pieces, 3-12 mm, 99.9% trace metals basis
Sigma-Aldrich
Zinc sulfide, purum, 97% (from Zn)
Sigma-Aldrich
Zinc sulfide, powder, 10 μm, 99.99% trace metals basis