Merck
  • [Effect of fluoride on expression of CaN mRNA and protein in bone tissue of rats].
[Effect of fluoride on expression of CaN mRNA and protein in bone tissue of rats].
Zhonghua bing li xue za zhi = Chinese journal of pathology (2013-01-11)
Ying Xie, Yan-ni Yu, Liang-bin Wan, Xi-shan Chen
ABSTRACT
To investigate the changes of mRNA and protein expression of CaN in the bone of rats with chronic fluorosis, and the mechanism of skeletal fluorosis. Thirty-six SD rats were divided into three groups (12 in each group, half male and half female selected according to body weight): control, low-dose and high-dose fluorosis groups. Controls were fed tap water (NaF < 0.5 mg/L), experimental animals in the low- or high-dose groups were fed water containing NaF of 5.0 and 50.0 mg/L, respectively. The rats were sacrificed after 6 months of treatment with fluoride. The serum was kept for testing bone metabolic marker bone gla protein (BGP) by enzyme-linked immunosorbent assay (ELISA), the protein and mRNA levels of CaN in distal femur of the rats with chronic flurosis were assessed by immunohistochemistry and in-situ hybridization. The levels of BGP (1.99 ± 0.62, 2.38 ± 0.16)µg/L in the low- or high-dose fluorosis groups were higher than that in the control group (0.15 ± 0.03) µg/L; and the high fluorosis group showed higher level than the low fluorosis group (all P < 0.05). Compared to the control group (131.11 ± 1.95, 111.82 ± 2.39), the protein and mRNA levels of CaN were higher in the low- or high-dose fluorosis groups (142.69 ± 1.17, 157.54 ± 1.88 and 121.28 ± 3.27, 134.63 ± 3.19, respectively), and the high fluorosis group showed higher levels than the low fluorosis group (all P < 0.05). BGP content could be used as a bone metabolic index in endemic fluorosis disease. Fluoride might up-regulate the mRNA and protein expression of CaN, and the changes in CaN level may be involved in the increase of the bone turnover and could be one of the pathogenetic factors in fluorosis.
Sigma-Aldrich
Sodium Fluoride Solution
Sigma-Aldrich
Sodium fluoride 0.5 M solution
Sigma-Aldrich
Sodium fluoride, anhydrous, powder, 99.99% trace metals basis
Supelco
Fluoride ion solution for ISE, 0.1 M F-, analytical standard (for ion-selective electrodes)
Sigma-Aldrich
Sodium fluoride, 99.99% trace metals basis
Sigma-Aldrich
Sodium fluoride, BioXtra, ≥99%
Sigma-Aldrich
Sodium fluoride, BioReagent, suitable for insect cell culture, ≥99%
Sigma-Aldrich
Sodium fluoride, JIS special grade, ≥99.0%
Sigma-Aldrich
Sodium fluoride, ReagentPlus®, ≥99%
Sigma-Aldrich
Sodium fluoride, ACS reagent, ≥99%
Sigma-Aldrich
Sodium fluoride, puriss., meets analytical specification of Ph. Eur., BP, USP, 98.5-100.5% (calc. to the dried substance)
Sigma-Aldrich
Fluoride standard solution, 1 mg/mL F-