Merck
  • Brain endogenous liver X receptor ligands selectively promote midbrain neurogenesis.
Brain endogenous liver X receptor ligands selectively promote midbrain neurogenesis.
Nature chemical biology (2013-01-08)
Spyridon Theofilopoulos, Yuqin Wang, Satish Srinivas Kitambi, Paola Sacchetti, Kyle M Sousa, Karl Bodin, Jayne Kirk, Carmen Saltó, Magnus Gustafsson, Enrique M Toledo, Kersti Karu, Jan-Åke Gustafsson, Knut R Steffensen, Patrik Ernfors, Jan Sjövall, William J Griffiths, Ernest Arenas
ABSTRACT
Liver X receptors (Lxrα and Lxrβ) are ligand-dependent nuclear receptors critical for ventral midbrain neurogenesis in vivo. However, no endogenous midbrain Lxr ligand has so far been identified. Here we used LC/MS and functional assays to identify cholic acid as a new Lxr ligand. Moreover, 24(S),25-epoxycholesterol (24,25-EC) was found to be the most potent and abundant Lxr ligand in the developing mouse midbrain. Both Lxr ligands promoted neural development in an Lxr-dependent manner in zebrafish in vivo. Notably, each ligand selectively regulated the development of distinct midbrain neuronal populations. Whereas cholic acid increased survival and neurogenesis of Brn3a-positive red nucleus neurons, 24,25-EC promoted dopaminergic neurogenesis. These results identify an entirely new class of highly selective and cell type-specific regulators of neurogenesis and neuronal survival. Moreover, 24,25-EC promoted dopaminergic differentiation of embryonic stem cells, suggesting that Lxr ligands may thus contribute to the development of cell replacement and regenerative therapies for Parkinson's disease.
Product Description
Cholic acid, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Cholic acid, from bovine and/or ovine, ≥98%