Merck
  • An open clinical trial on the efficacy of cetirizine hydrochloride in the management of allergic pruritus in cats.
An open clinical trial on the efficacy of cetirizine hydrochloride in the management of allergic pruritus in cats.
The Canadian veterinary journal = La revue veterinaire canadienne (2012-07-04)
Joya S Griffin, Danny W Scott, William H Miller, Michelle M Tranchina
ABSTRACT
Cetirizine hydrochloride was administered orally at 5 mg/cat, q24h, to 32 cats with allergic skin disease. Pruritus was reduced in 41% (13/32) of the cats. The antipruritic effect was repeatable and sustainable. There was no significant association between patient age, disease severity, or cutaneous reaction pattern and improvement during cetirizine administration. No adverse side effects were reported.
Sigma-Aldrich
Cetirizin dihydrochloride, ≥98.0% (HPLC)
Sigma-Aldrich
Cetirizine dihydrochloride, ≥98% (HPLC)
Cetirizine dihydrochloride, European Pharmacopoeia (EP) Reference Standard
Cetirizine for peak identification, European Pharmacopoeia (EP) Reference Standard