Merck
Luminescent iridium(III) complexes as novel protein staining agents.
Electrophoresis (2012-06-01)
Junli Jia, Hao Fei, Ming Zhou
ABSTRACT
This article reports a new class of luminescent metal complexes, biscyclometalated iridium(III) complexes with an ancillary bathophenanthroline disulfonate ligand, for staining protein bands that are separated by electrophoresis. The performances of these novel staining agents have been studied in comparison with tris(bathophenanthroline disulfonate) ruthenium(II) tetrasodium salt (i.e. RuBPS) using a commercially available imaging system. The staining agents showed different limits of detection, linear dynamic ranges, and protein-to-protein variations. The overall performances of all three stains were found to be better than or equivalent to RuBPS under the experimental conditions.
Sigma-Aldrich
Bathophenanthrolinedisulfonic acid disodium salt hydrate, ≥95%
Sigma-Aldrich
Bathophenanthrolinedisulfonic acid disodium salt hydrate, 98%
Supelco
Bathophenanthrolinedisulfonic acid disodium salt trihydrate, for the spectrophotometric det. of Fe, ≥98.0% (HPLC)