Merck
  • Engineering of a tyrosol-producing pathway, utilizing simple sugar and the central metabolic tyrosine, in Escherichia coli.
Engineering of a tyrosol-producing pathway, utilizing simple sugar and the central metabolic tyrosine, in Escherichia coli.
Journal of agricultural and food chemistry (2012-01-10)
Yasuharu Satoh, Kenji Tajima, Masanobu Munekata, Jay D Keasling, Taek Soon Lee
ABSTRACT
Metabolic engineering was applied to the development of Escherichia coli capable of synthesizing tyrosol (2-(4-hydroxyphenyl)ethanol), an attractive phenolic compound with great industrial value, from glucose, a renewable carbon source. In this strain, tyrosine, which was supplied not only from the culture medium but also from the central metabolism, was converted into tyrosol via three steps: decarboxylation, amine oxidation, and reduction. The engineered strain synthesized both tyrosol and 4-hydroxyphenylacetate (4HPA), but disruption of the endogenous phenylacetaldehyde dehydrogenase gene shut off 4HPA production and improved the production of tyrosol as a sole product. The engineered mutant strain was capable of producing 0.5 mM tyrosol from 1% (w/v) glucose during a 48 h shake flask cultivation.
Sigma-Aldrich
2-(4-Hydroxyphenyl)ethanol, 98%
Supelco
2-(4-Hydroxyphenyl)ethanol, analytical standard