Merck
  • Phthalate metabolites in urine from China, and implications for human exposures.
Phthalate metabolites in urine from China, and implications for human exposures.
Environment international (2011-04-12)
Ying Guo, Qian Wu, Kurunthachalam Kannan
ABSTRACT
Phthalates are esters of phthalic acid and are mainly used as plasticizers (added to plastics to increase their flexibility, transparency, durability, and longevity). Humans are exposed to phthalates through several routes. Urinary phthalate metabolites can be used as biomarkers of human exposures to phthalates. In this study, 14 phthalate metabolites were analyzed in 183 urine samples collected in 2010 from Shanghai, Guangzhou, and Qiqihaer, China. Phthalate metabolites were found in all urine samples and their total concentrations ranged from 18.6 to 3160 ng/mL (median: 331 ng/mL). Mono-n-butyl phthalate (mBP) and mono-2-isobutyl phthalate (miBP) were the major metabolites found in urine, and their respective median concentrations were 61.2 and 51.7 ng/mL; concentrations of miBP were higher than the concentrations reported for other countries, to date. Based on the urinary concentrations of phthalate metabolites, we estimated the daily intake rates in the Chinese population. The estimated daily intakes of dibutyl phthalate (DBP), diethyl phthalate (DEP), and di-(2-ethylhexyl) phthalate (DEHP) in China were 12.2, 3.8, and ~5 μg/kg bw/day, respectively. Thirty nine percent of the samples exceeded the tolerable daily intake of 10 μg/kg bw/day, proposed for DBP, by the European Food Safety Authority, but none of the estimated daily intake values exceeded the reference dose recommended by the U.S. Environmental Protection Agency.
Sigma-Aldrich
mono-Butyl phthalate, 97.0-103.0% (T)
Sigma-Aldrich
Diethyl phthalate, 99.5%
Sigma-Aldrich
Diethyl phthalate, ≥99%
Supelco
Diethyl phthalate, PESTANAL®, analytical standard
Sigma-Aldrich
Diethyl phthalate, SAJ special grade, ≥98.0%
Sigma-Aldrich
Diethyl phthalate, Technipur®, for synthesis