Merck
  • Protective effects of octacosanol on 6-hydroxydopamine-induced Parkinsonism in rats via regulation of ProNGF and NGF signaling.
Protective effects of octacosanol on 6-hydroxydopamine-induced Parkinsonism in rats via regulation of ProNGF and NGF signaling.
Acta pharmacologica Sinica (2010-06-29)
Tao Wang, Yan-yong Liu, Xin Wang, Nan Yang, Hai-bo Zhu, Ping-ping Zuo
ABSTRACT
To investigate the protective effects of octacosanol in 6-hydroxydopamine-induced Parkinsonian rats and find whether octacosanol has effects on pro nerve growth factor (pro-NGF), NGF and the downstream effector proteins. Behavioral tests, enzymatic assay, tyrosine hydroxylase immunohistochemistry, TUNEL and Western blot were used to investigate the effects of octacosanol in this rat model of PD. Oral administration of octacosanol (35-70 mg/kg, po for 14 d) significantly improved the behavioral impairments in rats induced by 6-OHDA and dose-dependently preserved the free radical scavenging capability of the striatum. Octacosanol treatment also effectively ameliorated morphological appearances of TH-positive neuronal cells in nigrostriatal systems and decreased the apoptotic cells induced by 6-OHDA in striatum. In addition, octacosanol strikingly blocked the 6-OHDA-induced increased expression of proNGF-p75NTR-sortilin death signaling complex and its downstream effector proteins. Meantime, octacosanol prevented the decreased levels of NGF, its receptors TrkA and p-Akt which together mediated the cell survival pathway. The findings implicated that the anti-parkinsonism effects afforded by octacosanol might be mediated by its neuro-microenvironment improving potency through retrieving the ratios of proNGF:NGF and the respective receptors p75NTR:TrkA in vivo. Due to its excellent tolerability and non-toxicity, octacosanol may be a promising agent for PD treatment.
Sigma-Aldrich
1-Octacosanol, ≥99% (GC)
Pricing and availability is not currently available.