Merck
  • Light intensity and light source influence on secondary organic aerosol formation for the m-xylene/NO(x) photooxidation system.
Light intensity and light source influence on secondary organic aerosol formation for the m-xylene/NO(x) photooxidation system.
Environmental science & technology (2008-08-30)
Bethany Warren, Chen Song, David R Cocker
ABSTRACT
A series of m-xylene/NO(x) photooxidation experiments were conducted to determine the influence of light intensity and radiation spectrum on secondary organic aerosol (SOA) formation within the UC Riverside/CE-CERT environmental chamber. The environmental chamber is equipped with 80 115-W black lights and a variable voltage 200 kW argon arc lamp that emits a wavelength spectrum more similar to natural light. SOA formation increased significantly with light intensity, measured as the photolysis rate of NO2 to NO (k1), increased from 0.09 to 0.26 min(-1). The argon arc lamp produced approximately 20% more SOA than black lights at a k1 of 0.09 min(-1) for similar amounts of m-xylene consumed. These results may help explain the variation of SOA formation between environmental chambers and the differences between measured SOA in the ambient atmosphere versus environmental chamber predictions.
Product Description
Supelco
m-Xylene, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
m-Xylene, anhydrous, ≥99%
Supelco
m-Xylene, analytical standard
Sigma-Aldrich
m-Xylene, puriss. p.a., ≥99.0% (GC)
Sigma-Aldrich
m-Xylene, ReagentPlus®, 99%