Merck
  • Effects of fluoride and ethanol administration on lipid peroxidation systems in rat brain.
Effects of fluoride and ethanol administration on lipid peroxidation systems in rat brain.
Indian journal of experimental biology (2013-05-18)
Shailender Singh Chauhan, Sudarshan Ojha, Akhtar Mahmood
ABSTRACT
Exposure to fluoride and excessive ethanol consumption has been identified as a serious public health problem in many parts of the world, including India. Thus, the effect of co-exposure to fluoride and ethanol for 3-6 weeks was studied on lipid peroxidation (LPO) and oxidative stress related parameters in the rat brain. After 3 weeks, co-treated animals showed 95% increase in LPO levels compared to control. However, the levels of reduced glutathione, total and protein thiols were decreased. These changes were accompanied by a decrease in the activities of superoxide dismutase, catalase, glutathione peroxidase, glutathione reductase and glutathione-S-transferase. Rats exposed to fluoride together with ethanol for 6 weeks resulted in 130% increase in LPO and decrease in the reduced glutathione levels. The activities of superoxide dismutase, catalase, glutathione peroxidase, glutathione reductase and glutathione-S-transferase were reduced under these conditions. Brain histology revealed excessive lymphocytes, edema and spongeosis in the cortical region after six weeks of fluoride and ethanol treatment. These results suggest that exposure to fluoride together with ethanol enhances lipid peroxidation by affecting antioxidant defence systems in the rat brain.
Sigma-Aldrich
Sodium Fluoride Solution
Sigma-Aldrich
Glutathione Reductase human, buffered aqueous solution, ≥10 units/mg protein, recombinant, expressed in E. coli
Sigma-Aldrich
Sodium fluoride, JIS special grade, ≥99.0%
Sigma-Aldrich
Fluoride standard solution, 1 mg/mL F-
Sigma-Aldrich
Sodium fluoride, anhydrous, powder, 99.99% trace metals basis
Supelco
Fluoride ion solution for ISE, 0.1 M F-, analytical standard (for ion-selective electrodes)
Sigma-Aldrich
Sodium fluoride, 99.99% trace metals basis
Sigma-Aldrich
Sodium fluoride, BioReagent, suitable for insect cell culture, ≥99%
Sigma-Aldrich
Glutathione Peroxidase from bovine erythrocytes, lyophilized powder, ≥300 units/mg protein
Sigma-Aldrich
Sodium fluoride, BioXtra, ≥99%
Sigma-Aldrich
Glutathione S-Transferase from equine liver, lyophilized powder, ≥25 units/mg protein
Sigma-Aldrich
Sodium fluoride 0.5 M solution
Sigma-Aldrich
Sodium fluoride, ReagentPlus®, ≥99%
Sigma-Aldrich
Sodium fluoride, puriss., meets analytical specification of Ph. Eur., BP, USP, 98.5-100.5% (calc. to the dried substance)
Sigma-Aldrich
Sodium fluoride, ACS reagent, ≥99%