Merck
  • Spatiotemporal refinement of signal flow through association cortex during learning.
Spatiotemporal refinement of signal flow through association cortex during learning.
Nature communications (2020-04-10)
Ariel Gilad, Fritjof Helmchen
ABSTRACT
Association areas in neocortex encode novel stimulus-outcome relationships, but the principles of their engagement during task learning remain elusive. Using chronic wide-field calcium imaging, we reveal two phases of spatiotemporal refinement of layer 2/3 cortical activity in mice learning whisker-based texture discrimination in the dark. Even before mice reach learning threshold, association cortex-including rostro-lateral (RL), posteromedial (PM), and retrosplenial dorsal (RD) areas-is generally suppressed early during trials (between auditory start cue and whisker-texture touch). As learning proceeds, a spatiotemporal activation sequence builds up, spreading from auditory areas to RL immediately before texture touch (whereas PM and RD remain suppressed) and continuing into barrel cortex, which eventually efficiently discriminates between textures. Additional correlation analysis substantiates this diverging learning-related refinement within association cortex. Our results indicate that a pre-learning phase of general suppression in association cortex precedes a learning-related phase of task-specific signal flow enhancement.
Sigma-Aldrich
Thymidine 5′-monophosphate disodium salt hydrate, ≥99%
Pricing and availability is not currently available.
Sigma-Aldrich
Trimethoprim, ≥98.5%
Pricing and availability is not currently available.
Sigma-Aldrich
Dimethyl sulfoxide, suitable for HPLC, ≥99.7%
Pricing and availability is not currently available.