
Testosterone-d3 (16, 16, 17-d3) solution

Synonym(s):
Testosterone-d3 solution
CAS Number:
EC Number:
Products
Product Category
Brand
Manufacturer
Physical Form
Format
Quality Segment
Application
Package Size
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing