Skip to Content
Merck
Protoporphyrin IX disodium salt ≥90% (HPLC)

Protoporphyrin IX disodium salt

Protoporphyrin IX disodium salt ≥90% (HPLC)
Synonym(s):
3,7,12,17-Tetramethyl-8,13-divinyl-2,18-porphinedipropionic acid disodium salt, Depocolin-S, Ooporphyrin disodium salt
Empirical Formula (Hill Notation):
C34H32N4Na2O4
CAS Number:
Molecular Weight:
606.62
EC Number:

Products

Product Category
Brand
Formula Weight
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing