
Inosine 5′-monophosphate disodium salt hydrate

Synonym(s):
5′-IMP-Na2, 5′-Inosinic acid disodium salt hydrate, I-5′-P, IMP, Inosinic Acid
Empirical Formula (Hill Notation):
C10H11N4O8PNa2 · xH2O
CAS Number:
Molecular Weight:
392.17 (anhydrous basis)
Products
Brand
Biological Source
Formula Weight
Physical Form
Purity
Showing 1-2 of 2
Compare
Product Number
Description
Pricing