
Fast β-Glucuronidase, Recombinant

Synonym(s):
β-Glucuronidase from limpets (Patella vulgata), β-D-Glucuronide glucuronosohydrolase
CAS Number:
Products
Product Category
Brand
Physical Form
Recombinant Host
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing