
Ethanol-500

Synonym(s):
Ethanol solution, Ethyl alcohol
Linear Formula:
C2H5OH
CAS Number:
Molecular Weight:
46.07
Products
Product Category
Brand
Formula Weight
Manufacturer
Physical Form
Format
Quality Segment
Application
Package Size
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing