
Clonazepam-d4 solution

Synonym(s):
Clonazepam-d4 solution
Empirical Formula (Hill Notation):
C15D4H6ClN3O3
Products
Brand
Formula Weight
Manufacturer
Physical Form
Format
Application
Package Size
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2