Merck
Posttranslational arginylation enzyme Ate1 affects DNA mutagenesis by regulating stress response.
Cell death & disease (2016-09-30)
Akhilesh Kumar, Michael D Birnbaum, Devang M Patel, William M Morgan, Jayanti Singh, Antoni Barrientos, Fangliang Zhang
RESUMEN
Arginyltransferase 1 (Ate1) mediates protein arginylation, a poorly understood protein posttranslational modification (PTM) in eukaryotic cells. Previous evidence suggest a potential involvement of arginylation in stress response and this PTM was traditionally considered anti-apoptotic based on the studies of individual substrates. However, here we found that arginylation promotes cell death and/or growth arrest, depending on the nature and intensity of the stressing factor. Specifically, in yeast, mouse and human cells, deletion or downregulation of the ATE1 gene disrupts typical stress responses by bypassing growth arrest and suppressing cell death events in the presence of disease-related stressing factors, including oxidative, heat, and osmotic stresses, as well as the exposure to heavy metals or radiation. Conversely, in wild-type cells responding to stress, there is an increase of cellular Ate1 protein level and arginylation activity. Furthermore, the increase of Ate1 protein directly promotes cell death in a manner dependent on its arginylation activity. Finally, we found Ate1 to be required to suppress mutation frequency in yeast and mammalian cells during DNA-damaging conditions such as ultraviolet irradiation. Our study clarifies the role of Ate1/arginylation in stress response and provides a new mechanism to explain the link between Ate1 and a variety of diseases including cancer. This is also the first example that the modulation of the global level of a PTM is capable of affecting DNA mutagenesis.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Roche
Kit de detección de muerte celular in situ, fluoresceína, sufficient for ≤50 tests, suitable for detection
Sigma-Aldrich
Anti-β-actina, anticuerpo monoclonal, clone AC-15, purified from hybridoma cell culture
Sigma-Aldrich
Peróxido de hidrógeno solution, contains ~200 ppm acetanilide as stabilizer, 3 wt. % in H2O
Sigma-Aldrich
Mechlorethamine hydrochloride, 98%
Sigma-Aldrich
Anti-ATE1 Antibody, clone 6F11, clone 6F11, from rat