Merck
The first radiosynthesis of [(11)C]AZD8931 as a new potential PET agent for imaging of EGFR, HER2 and HER3 signaling.
Bioorganic & medicinal chemistry letters (2014-08-26)
Min Wang, Mingzhang Gao, Qi-Huang Zheng
RESUMEN
The reference standard AZD8931{2-(4-((4-((3-chloro-2-fluorophenyl)amino)-7-methoxyquinazolin-6-yl)oxy)piperidin-1-yl)-N-methylacetamide} (11a) was synthesized from methyl 4,5-dimethoxy-2-nitrobenzoate or ethyl 4,5-dimethoxy-2-nitrobenzoate and 2-chloro-N-methylacetamide in 11 steps with 2-5% overall chemical yield. The precursor N-desmethyl-AZD8931{2-(4-((4-((3-chloro-2-fluorophenyl)amino)-7-methoxyquinazolin-6-yl)oxy)piperidin-1-yl)acetamide} (11b) was synthesized from methyl 4,5-dimethoxy-2-nitrobenzoate or ethyl 4,5-dimethoxy-2-nitrobenzoate and 2-bromoacetamide in 11 steps with 2-4% overall chemical yield. The target tracer [(11)C]AZD8931 {2-(4-((4-((3-chloro-2-fluorophenyl)amino)-7-methoxyquinazolin-6-yl)oxy)piperidin-1-yl)-N-[(11)C]methylacetamide} ([(11)C]11a) was prepared from N-desmethyl-AZD8931 (11b) with [(11)C]CH3OTf under basic condition (NaH) through N-[(11)C]methylation and isolated by HPLC combined with solid-phase extraction (SPE) in 40-50% radiochemical yield based on [(11)C]CO2 and decay corrected to end of bombardment (EOB) with 370-1110GBq/μmol specific activity at EOB.