Merck
  • Prognostic utility of plasma S100A12 levels to establish a novel scoring system for predicting mortality in maintenance hemodialysis patients: a two-year prospective observational study in Japan.
Prognostic utility of plasma S100A12 levels to establish a novel scoring system for predicting mortality in maintenance hemodialysis patients: a two-year prospective observational study in Japan.
BMC nephrology (2013-01-18)
Yayoi Shiotsu, Yasukiyo Mori, Masato Nishimura, Tsuguru Hatta, Naoki Imada, Noboru Maki, Kumiko Iida, Noriyuki Iwamoto, Eiko Matsuoka, Keiichi Tamagaki, Atsushi Kosaki
RESUMEN
S100A12 protein is an endogenous receptor ligand for advanced glycation end products. In this study, the plasma S100A12 level was assessed as an independent predictor of mortality, and its utility in clinical settings was examined. In a previous cross-sectional study, plasma S100A12 levels were measured in 550 maintenance hemodialysis patients to determine the association between S100A12 and the prevalence of cardiovascular diseases (CVD). In this prospective study, the risk of mortality within a two-year period was determined. An integer scoring system was developed to predict mortality on the basis of the plasma S100A12 levels. Higher plasma S100A12 levels (≥18.79 ng/mL) were more closely associated with higher all-cause mortality than lower plasma S100A12 levels (<18.79 ng/mL; P = 0.001). Multivariate Cox proportional hazards analysis revealed higher plasma S100A12 levels [hazard ratio (HR), 2.267; 95% confidence interval (CI), 1.195-4.302; P = 0.012], age ≥65 years (HR, 1.961; 95%CI, 1.017-3.781; P = 0.044), serum albumin levels <3.5 g/dL (HR, 2.198; 95%CI, 1.218-3.968; P = 0.012), and history of CVD (HR, 2.068; 95%CI, 1.146-3.732; P = 0.016) to be independent predictors of two-year all-cause mortality. The integer score was derived by assigning points to these factors and determining total scores. The scoring system revealed trends across increasing scores for predicting the all-cause mortality [c-statistic = 0.730 (0.656-0.804)]. The resulting model demonstrated good discriminative power for distinguishing the validation population of 303 hemodialysis patients [c-statistic = 0.721 (0.627-0.815)]. The results indicate that plasma S100A12 level is an independent predictor for two-year all-cause mortality. A simple integer scoring system was therefore established for predicting mortality on the basis of plasma S100A12 levels.