Merck
  • Silica coating and photochemical properties of layered double hydroxide/4,4'-diaminostilbene-2,2'-disulfonic acid nanocomposite.
Silica coating and photochemical properties of layered double hydroxide/4,4'-diaminostilbene-2,2'-disulfonic acid nanocomposite.
Journal of colloid and interface science (2005-07-19)
Ahmed Mohamed El-Toni, Shu Yin, Tsugio Sato
RESUMEN
Organic ultraviolet (UV) rays absorbents have been used as sunscreen materials, but they may pose a safety problem when used at high concentration. In order to prevent direct contact of organic UV rays absorbent to the human skin, a typical organic UV-absorbent, 4,4(')-diaminostilbene-2,2(')-disulfonic acid (DASDSA), was intercalated into Zn(2)Al layered double hydroxide (Zn(2)Al-LDH) by coprecipitation reaction. However, deintercalation of DASDSA from Zn(2)Al-LDH, by the anion exchange reaction with carbonate ion, was observed. Therefore, Zn(2)Al-LDH/DASDSA was directly coated with silica by means of polymerization technique based on the Stöber method. Silica coating effectively depressed the deintercalation of DASDSA from Zn(2)Al-LDH. The amorphous silica was confirmed by XRD, SEM, TEM and FT-IR. The deintercalation behaviors as well as UV-shielding properties were investigated for coated particles.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
4,4′-Diamino-2,2′-stilbenedisulfonic acid, technical grade, 85%