Merck
Role of TAZ as mediator of Wnt signaling.
Cell (2012-12-19)
Luca Azzolin, Francesca Zanconato, Silvia Bresolin, Mattia Forcato, Giuseppe Basso, Silvio Bicciato, Michelangelo Cordenonsi, Stefano Piccolo
RESUMEN
Wnt growth factors are fundamental regulators of cell fate, but how the Wnt signal is translated into biological responses is incompletely understood. Here, we report that TAZ, a biologically potent transcriptional coactivator, serves as a downstream element of the Wnt/β-catenin cascade. This function of TAZ is independent from its well-established role as mediator of Hippo signaling. In the absence of Wnt activity, the components of the β-catenin destruction complex--APC, Axin, and GSK3--are also required to keep TAZ at low levels. TAZ degradation depends on phosphorylated β-catenin that bridges TAZ to its ubiquitin ligase β-TrCP. Upon Wnt signaling, escape of β-catenin from the destruction complex impairs TAZ degradation and leads to concomitant accumulation of β-catenin and TAZ. At the genome-wide level, a substantial portion of Wnt transcriptional responses is mediated by TAZ. TAZ activation is a general feature of Wnt signaling and is functionally relevant to mediate Wnt biological effects.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Leukocyte Alkaline Phosphatase Kit, based on naphthol AS-MX phosphate and fast blue RR salt (with citrate)