Merck
Claudin-4 as therapeutic target in cancer.
Archives of biochemistry and biophysics (2012-01-31)
A Neesse, H Griesmann, T M Gress, P Michl
RESUMEN
Intercellular junctional complexes such as adherens junctions and tight junctions are critical regulators of cellular polarity, paracellular permeability and metabolic and structural integrity of cellular networks. Abundant expression analysis data have yielded insights into the complex pattern of differentially expressed cell-adhesion proteins in epithelial cancers and provide a useful platform for functional, preclinical and clinical evaluation of novel targets. This review will focus on the role of claudin-4, an integral constituent of tight junctions, in the pathophysiology of epithelial malignancies with particular focus pancreatic cancer, and its potential applicability for prognostic, diagnostic and therapeutic approaches. Claudin-4 expression is widely dysregulated in epithelial malignancies and in a number of premalignant precursor lesions. Although the functional implications are only starting to unravel, claudin-4 seems to play an important role in tumour cell invasion and metastasis, and its dual role as receptor of Clostridium perfringens enterotoxin (CPE) opens exciting avenues for molecular targeted approaches. Claudin-4 constitutes a promising molecular marker for prognosis, diagnosis and therapy of epithelial malignancies.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Claudin-4 (EP417) Rabbit Monoclonal Primary Antibody