Merck
Glycation potentiates α-synuclein-associated neurodegeneration in synucleinopathies.
Brain : a journal of neurology (2017-04-12)
Hugo Vicente Miranda, Éva M Szego, Luís M A Oliveira, Carlo Breda, Ekrem Darendelioglu, Rita M de Oliveira, Diana G Ferreira, Marcos A Gomes, Ruth Rott, Márcia Oliveira, Francesca Munari, Francisco J Enguita, Tânia Simões, Eva F Rodrigues, Michael Heinrich, Ivo C Martins, Irina Zamolo, Olaf Riess, Carlos Cordeiro, Ana Ponces-Freire, Hilal A Lashuel, Nuno C Santos, Luisa V Lopes, Wei Xiang, Thomas M Jovin, Deborah Penque, Simone Engelender, Markus Zweckstetter, Jochen Klucken, Flaviano Giorgini, Alexandre Quintas, Tiago F Outeiro
RESUMEN
α-Synuclein misfolding and aggregation is a hallmark in Parkinson's disease and in several other neurodegenerative diseases known as synucleinopathies. The toxic properties of α-synuclein are conserved from yeast to man, but the precise underpinnings of the cellular pathologies associated are still elusive, complicating the development of effective therapeutic strategies. Combining molecular genetics with target-based approaches, we established that glycation, an unavoidable age-associated post-translational modification, enhanced α-synuclein toxicity in vitro and in vivo, in Drosophila and in mice. Glycation affected primarily the N-terminal region of α-synuclein, reducing membrane binding, impaired the clearance of α-synuclein, and promoted the accumulation of toxic oligomers that impaired neuronal synaptic transmission. Strikingly, using glycation inhibitors, we demonstrated that normal clearance of α-synuclein was re-established, aggregation was reduced, and motor phenotypes in Drosophila were alleviated. Altogether, our study demonstrates glycation constitutes a novel drug target that can be explored in synucleinopathies as well as in other neurodegenerative conditions.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Anticuerpo anti-AGE (productos finales de glucosilación avanzada), serum, Chemicon®
Sigma-Aldrich
Anti-Aggregated a-Synuclein Antibody, clone 5G4, clone 5G4, from mouse, purified by affinity chromatography