Key Documents
Z758094
AC adapter for BRAND® PLT unit
pkg of 1 ea
About This Item
UNSPSC Code:
41121500
NACRES:
NB.01
packaging
pkg of 1 ea
manufacturer/tradename
BRAND 703979
General description
Input: AC 100-240 V; 50/60 Hz, Output: DC 6,5 V, 800 mA
Other Notes
BR703979 (India)
Legal Information
BRAND is a registered trademark of BRAND GMBH + CO KG
Optional